o-Phenylenediamine dihydrochloride ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Benzene-1,2-diamine dihydrochloride, 1,2-Phenylenediammonium dichloride, 1,2-Diaminobenzene dihydrochloride, 1,2-Phenylenediamine dihydrochloride

J60354.14 J60354.22
J60354.14EA 62.4 GBP
J60354.14 J60354.22
o-Phenylenediamine dihydrochloride ≥98%
o-Phenylenediamine dihydrochloride

Purity: 98+% CAS No.: 615-28-1 Molecular Formula: C6H8N2.2HCl Formula Weight: 181.07 Storage Temperature: -30°C to -10°C Physical Form: Powder Beilstein Reference: 3912045 UN No.: 2811 MDL No.: MFCD00012966


A peroxidase substrate for ELISA procedures

Formula: C₆H₁₀Cl₂N₂
MW: 181.06 g/mol
Melting Pt: 205
MDL Number: MFCD00012966
CAS Number: 615-28-1
EINECS: 210-418-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR