Glycine 99+%, Ultrapure for molecular biology

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

J16407.A1 J16407.A7 J16407.36
J16407.A1EA 99.8 GBP
J16407.A1 J16407.A7 J16407.36
Glycine 99+%, Ultrapure for molecular biology
Glycine
Formula: NH₂CH₂COOH
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: ∼ 245 °C (dec.)
Density: 1.595
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 14,04491
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Unspecified) ≥99.0%
Assay (Unspecified) - Hydrolysable Substances Passes Test
Assay (Unspecified) - Readily Carbonisable Substances Passes Test
Chloride Content ≤50 ppm
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Impurity Content Sulphate (SO₄) ≤50 ppm
lambda-max (UV-Vis) - (1 M, 1 cm ≤0.05) 260 nm
Arsenic (As) ≤1 ppm
Barium (Ba) ≤1 ppm
Cadmium (Cd) ≤1 ppm
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Manganese (Mn) ≤1 ppm
Zinc (Zn) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR