Anti-IgG Donkey Polyclonal Antibody (DyLight® 405)

Supplier: ImmunoReagents
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

IR2879
IMMRIR2879EA 121 GBP
IMMRIR2879
Anti-IgG Donkey Polyclonal Antibody (DyLight® 405)
Antibodies
The ImmunoReagents' DyLight 405 has a vibrant blue fluorescence intensity that replaces Alexa Fluor™ 405, Cascade™ Blue and AMCA in many applications. The high water solubility of DyLight Fluors means that a high dye-to-protein ratio can be attained without causing precipitation of the conjugates. DyLight® 405 (Ex = 400 nm; Em = 420 nm)

This conjugate is suitable for immunomicroscopy and flow cytometry. The optimal working dilution should be determined by the investigator. Suggested starting dilution(?): · 1:20 - 1:2,000 for most applications

Type: Secondary
Antigen: IgG
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR