Anti- Llama Polyclonal Antibody (Unconjugated)

Supplier: ImmunoReagents
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

IR2400
IMMRIR2400EA 38.7 GBP
IMMRIR2400
Anti- Llama Polyclonal Antibody (Unconjugated)
Antibodies
Type:
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Llama
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR