Anti-IgG Goat Polyclonal Antibody (DyLight® 800)
Supplier: ImmunoReagents
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
IR1455
IMMRIR1455EA 262 GBP
IMMRIR1455
Anti-IgG Goat Polyclonal Antibody (DyLight® 800)
Antibodies
The ImmunoReagents' DyLight® 800 conjugates uses a near-IR fluorphore that is invisible to the naked eye but increases the staining options when using infrared imaging systems. DyLight® 800 has spectral properties that are very similar to other near-IR dyes, including Alexa Fluor 790 and IRDye 800, and has high solubility for maximal incorporation of the dye into the biomolecule of interest. Excitation/Emission = 777 nm/794 nm Emission Colour = Near IR (Similar Dyes: Alexa Fluor 790, IRDye 800).
  • Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats andMore Product Information