Anti-IgG Chicken Polyclonal Antibody (Biotin (NHS-LC))
Supplier: ImmunoReagents
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
IR1049
IMMRIR1049EA 165 GBP
IMMRIR1049
Anti-IgG Chicken Polyclonal Antibody (Biotin (NHS-LC))
Antibodies
ImmunoReagents’ Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualised, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.
  • Origin: US
ImmunoReagents' antibodies areMore Product Information