tempmate.®-C1 Dry Ice Temperature Data Logger
Supplier: tempmate
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TC1-000
IMECTC1-000EA 97.9 GBP
IMECTC1-000
tempmate.®-C1 Dry Ice Temperature Data Logger
Data Loggers
Single-use dry-ice monitoring solution for direct measurements in ultra-low temperatures without accessories.
  • Pre-programmed
  • 90 days active runtime (restartable)
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food grade packing
  • No additional hardware required
  • Detailed PDF and CSV report including graph and table
  • Big LED Display for status monitoring
  • Extended shelf-life of two years
Your tempmate.®-C1 can record temperatures as low as −90 °C and thus even provide a reliable overview of your dry ice transports. For thisMore Product Information