Butterfly valves, VK series
Supplier: Welch by Gardner Denver
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
704523 704522
ILMV704523EA 398 GBP
ILMV704523 ILMV704522
Butterfly valves, VK series
Valves Vacuum Valves
These manually operated butterfly valves are made of stainless steel with viton seal.
Leak rate 10-7 mbar L/s