High vacuum angle valves, with sealed bellows, VA series
Supplier: Welch by Gardner Denver
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
700101 700371 700370 700109
ILMV700101EA 484 GBP
ILMV700101 ILMV700371 ILMV700370 ILMV700109
High vacuum angle valves, with sealed bellows, VA series
Valves Vacuum Valves
These high vacuum angle valves feature a very long life bellows-seal made from stainless steel.
  • High conductance
  • Leak rate 10-9 mbar L/s