Welch® CRVpro™ Rotary Vane Vacuum Pumps
Supplier: Welch by Gardner Denver
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3062-00 330072 3162-00 330071 3242-00 3022-00 3302-00 3042-00 3082-00 3087T-01 330070 330073 3487T-01 3657T-01 330079 330078
ILMV3062-00EA 2590 GBP
ILMV3062-00 ILMV330072 ILMV3162-00 ILMV330071 ILMV3242-00 ILMV3022-00 ILMV3302-00 ILMV3042-00 ILMV3082-00 ILMV3087T-01 ILMV330070 ILMV330073 ILMV3487T-01 ILMV3657T-01 ILMV330079 ILMV330078
Welch® CRVpro™ Rotary Vane Vacuum Pumps
Pumps Vacuum Pumps
These two-stage rotary vane vacuum pumps are designed to simplify task and provides highest reliability for stable operation and long product lifespan.
  • Low noise level
  • Anti-suck back valve
  • Forced oil lubrication
The motors of these pumps are dual voltage functional that is switchable from 115 to 240 V. Cool running operation of this pump provides less oil consumption and slowing down corrosion.
These provide a measure of protection from sublimed chemical vapors with coating on pump module and oil case. Ensures the extended usability of oil due toMore Product Information