Magnetic stirrer, Midi MR 1 digital IKAMAG®
Supplier: IKA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25002968 25004056
IKAA25002968EA 3490 GBP
IKAA25002968 IKAA25004056
Magnetic stirrer, Midi MR 1 digital IKAMAG®
Stirrers Magnetic Stirrers
Powerful high capacity magnetic stirrer with digital LED display.
  • Infinitely variable speed
  • Non-locking motor
  • Continuous operation or timer mode from 0 to 56 min
Plus models feature integrated USB and RS232 connections.
Certifications: IP Protection Class according to DIN EN 60529: IP 21.