ROTAVISC Viscometer, ROTATEMP SBS me-vi Package
Supplier: IKA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
10013995
IKAA10013995EA 0 GBP
IKAA10013995
ROTAVISC Viscometer, ROTATEMP SBS me-vi Package
Viscometers Rotational Viscometers
The ROTATEMP SBS me-vi package is the ideal solution for precise and reliable viscosity measurement of difficult-to-measure, highly viscous samples.
  • Fast, precise and reproducible results
  • The combination of ROTAVISC viscometer and ROTATEMP high-temperature system simplifies the viscosity measurement of difficult-to-measure, highly viscous samples
  • Simple operation and space-saving
  • The complete system is supplied ready for connection
  • With its space-saving design, it fits easily into any laboratory
  • Optimum compatibility and automation
  • ROTAVISC and ROTATEMP
More Product Information