Glass beads, MP Biomedicals
Supplier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
116914801
ICNA116914801EA 118 GBP
ICNA116914801
Glass beads, MP Biomedicals
Beads Glass Beads
Glass beads.
  • For research use only
  • Can be stored at 2 to 8 °C
Glass beads used as packing for distillation columns, mixing beads and boiling stones.