Dehydrated culture media, YNB media, MP Biomedicals
Supplier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
114027542 114027812 114027532 114027522 114027512 114027022 114027012 114027032 114027112 114027412 114029622 114030112
ICNA114027542EA 0 GBP
ICNA114027542 ICNA114027812 ICNA114027532 ICNA114027522 ICNA114027512 ICNA114027022 ICNA114027012 ICNA114027032 ICNA114027112 ICNA114027412 ICNA114029622 ICNA114030112
Dehydrated culture media, YNB media, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
YNB media, a component of synthetic defined yeast media is a well defined composition of salts, vitamins and nitrogen source for a vigorous growth of S. cerevisiae.