Dehydrated culture media, YPD (YEPD) media, MP Biomedicals
Supplier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
114001065 114001075 114001275
ICNA114001065EA 115 GBP
ICNA114001065 ICNA114001075 ICNA114001275
Dehydrated culture media, YPD (YEPD) media, MP Biomedicals
Media for Microbiology Prepared Media for Microbiology
YPD broth or agar are used for routine growth of Saccharomyces cerevisiae for molecular biology studies.
  • Available in ready-to-use pouches
  • YPD agar media contains agar in addition to the other ingredients
It is a nutritionally rich medium containing dextrose, peptone and yeast extract. Dextrose is the energy source. Yeast extract and peptone provide carbon, nitrogen, minerals, vitamins, trace ingredients and other essential growth nutrients which stimulate the growth of yeasts.