Corn oil (from corn), yellow oil

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0290141401 0290141405 0290141425
ICNA0290141401EA 51.8 GBP
ICNA0290141401 ICNA0290141405 ICNA0290141425
Corn oil (from corn), yellow oil
Corn oil

Corn oil is used as a research animal diet supplement. It is also used in research applications as delivery vehicle for fat soluble compounds including toxins.


Oleic acid: 43 - 47%, linoleic acid: 39 - 42%; palmitic acid: 7 - 8%; stearic acid: 3 - 4%; arachidic acid: 0,5%; approx. 7,6 lbs/gal (Lit.).

Melting Pt: -10
Storage Temperature: Ambient
MDL Number: MFCD00130865
CAS Number: 8001-30-7

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR