Eosin B spirit soluble free acid ∼ 85% (dye content), powder C.I. 45400

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0219516410
ICNA0219516410EA 70.1 GBP
ICNA0219516410
Eosin B spirit soluble free acid ∼ 85% (dye content), powder C.I. 45400
Eosin B spirit soluble

Soluble in heated methanol (1 mg/ml)

Formula: C20H8Br2N2O9
Melting Pt: 275 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD01104119
CAS Number: 56360-46-4

Order Now

Specification Test Results

UV/Vis: λ max, methanol 522 ± 2 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR