Thiamine hydrochloride, powder

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

0219474983 0219474925 0219474980
ICNA0219474983EA 263 GBP
ICNA0219474983 ICNA0219474925 ICNA0219474980
Thiamine hydrochloride, powder
Thiamine hydrochloride

Used in fluorometric determination of mercury. Required to support energy metabolism and amino acid synthesis in cultured cells. Present in many classical and serum-free formulations.


Caution: Research use only (RUO). Ready for CE IVD labeling of clinical applications.

Formula: C₁₂H₁₈Cl₂N₄OS
Melting Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00012780
CAS Number: 67-03-8
EINECS: 200-641-8

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
pH 2,7 - 3,4 (1% aq soln)
UV/VIS: OD400 ≤0,25 (1% aq soln)
Loss on Drying ≤5,0%
Residue on Ignition ≤0,2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR