Protamine sulphate (from salmon sperm), ≥100 USP u/mg, white powder cell culture reagent

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Salmine sulphate

0219472925 0219472980 0219472901 0219472905
ICNA0219472925EA 267 GBP
ICNA0219472925 ICNA0219472980 ICNA0219472901 ICNA0219472905
Protamine sulphate (from salmon sperm), ≥100 USP u/mg, white powder cell culture reagent
Protamine sulphate

Storage: Room Temperature, desiccate
Protamine sulfate is a purified mixture of simple protein principles obtained from the sperm or testes of suitable species of fish, which has the property of neutralizing heparin. Because of having many basic amino acids (mostly arginine) protamine contains far greater nitrogen than other proteins. Its molecular weight is relatively small. Histone and other basic proteins in the testes of unmatured fishes convert into protamine along with the growth of the fishes. In the testes, protamine takes the form of nucleoprotamine linked with DNA.
Protamine Sulfate is a raw material for study preparations like insulin compounds, and etc. It is used in separation and refining of vaccines. It is a reagent for removing nucleic acids from enzyme solution for the purpose of easy separation and refining. Protamine in the form of solid lipid nanoparticles (SLN) promoted transfection with plasmid DNA more efficiently and with less cytotoxicity than comparable SLNs composed of Esterquat-1.
Protamine sulfate is a small cationic protein that binds and precipitates DNA. Inhibits lipoprotein lipase. Protamine sulfate shown to inhibit the classical pathway of complement. It inhibits turnover of lipoproteins by lipoprotein lipase.


  • Cell Culture Reagent Grade
  • Presentation: White Powder
  • Freely soluble in hot water, slightly soluble in cold water; soluble in 0.1 M KH2PO4; Soluble in HCl. Practically insoluble in organic solvents.

Storage Temperature: Ambient
MDL Number: MFCD00132091
CAS Number: 9009-65-8

Order Now

Specification Test Results

Identity Test Passes
Source Salmon Sperm
Loss on Drying ≤5%
Sulfate 16-22%
Nitrogen 22.5-25.5%
Activity ≥100 USP u/mg

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR