Sodium-L(+)-glutamate monohydrate ≥99%, white powder cell culture reagent

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0219467780 0219467790 0219467791
ICNA0219467780EA 41.2 GBP
ICNA0219467780 ICNA0219467790 ICNA0219467791
Sodium-L(+)-glutamate monohydrate ≥99%, white powder cell culture reagent
Sodium-L(+)-glutamate monohydrate

L-Glutamic Acid is a non-essential amino acid for human development; referred to as an excitatory amino acid (EAA) due to its role in neurotransmission. Acts as a neurotransmitter at fast synapses; agonist at kainate, NMDA and quisqualate receptors. L-Glutamic acid is one of the two amino acids that contains a carboxylic acid group in its side chains.


L-Glutamic acid is used in cell culture as a component of MEM non-essential amino acids solution.
Agonist at kainate, NMDA, and quisqualate receptors; an excitatory amino acid neurotransmitter.

Formula: C₅H₈NNaO₄·H₂O
Melting Pt: 232 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00150138
CAS Number: 6106-04-3

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
Optical Rotation (c=1, 1 N HCl) +20° to +36°
Loss on Drying <0.5%
pH (5%, aq soln) 6.7-7.2
Solubility (10% aq soln) Clear and Colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR