Bovine aprotinin (from Lung), MP Biomedicals

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0219115801 0219115805 0219115810 0219115825 0219115880
ICNA0219115801EA 38.4 GBP
ICNA0219115801 ICNA0219115805 ICNA0219115810 ICNA0219115825 ICNA0219115880
Bovine aprotinin (from Lung), MP Biomedicals
Proteins and Peptides

Aprotinin is a protein consisting of 58 amino acids, arranged in a single polypeptide chain that is cross-linked by three disulphide bridges. It is found in bovine lymph nodes, lung, parotid gland, spleen, liver, pancreas, seminal vesicles, thyroid gland, kidney, mucous membranes of the trachea and esophagus, ovaries, heart, posterior pituitary and cartilage.


  • Extinction Coefficient (E1%)(280 nm) = 8,3 (water)
  • Isoelectric point (pI): 10,5
  • Soluble in water and in aqueous buffers of low ionic strengths
  • Sterilisation of solutions can be done by filtration through a 0,2 µm filter


It is a competitive serine protease inhibitor that inhibits trypsin, chymotrypsin, kallikrein and plasmin. It forms stable complexes with and blocks the active sites of enzymes. Binding is reversible with most aprotinin-protease complexes dissociating at pH >10 or < 3.


Unit definition: The quantity of protease inhibitor that has the ability to inhibit 2 Kallikrein units by 50% under optimal conditions.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR