Digitonin, white powder

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Digitonin

0215948082 0215948090
ICNA0215948082EA 191 GBP
ICNA0215948082 ICNA0215948090
Digitonin, white powder
Digitonin

Digitonin is a mild nonionic detergent. It permeabilizes plasma membranes of eukaryotic cells by complexing with membrane cholesterol and other unconjugated β-hydroxysterols.


  • Water soluble
  • Critical Micelle Concentration (CMC): <0.5 mM (20…25 °C)(Lit.)
  • [α]20/D −54 °, c = 2.8 in methanol (Lit.)


Digitonin has used for cholesterol determination, use of digitonin-permeabilised adipocytes for cAMP studies, solubilisation of membrane proteins. The water soluble form can be used for the isolation of mitotic chromosomes. Mild nonionic detergent used to solubilise receptors and permeabilise cellular and nuclear membranes.


Soluble in boiling water (1g/10 ml). Heat for 15 minutes at 98 °C. Cool and place at 4 °C for 3 hrs and filter off any precipitate. Soluble in absolute alcohol (1 g / 57 ml) and 95% alcohol (1 g).

Formula: C₅₆H₉₂O₂₉
Melting Pt: 230…240 °C (dec., lit.)
MDL Number: MFCD00077729
CAS Number: 11024-24-1
EINECS: 234-255-6
UN: 3462
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR