Lithium sulphate monohydrate ≥99.0% (dried basis), white powder ACS

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

0215253990 0215253980
ICNA0215253990EA 347 GBP
ICNA0215253990 ICNA0215253980
Lithium sulphate monohydrate ≥99.0% (dried basis), white powder ACS
Lithium sulphate monohydrate

Soluble in cold water.

Formula: Li₂SO₄·H₂O
Melting Pt: 120 °C
Density: 2.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149766
CAS Number: 10102-25-7
EINECS: 233-820-4

Order Now

Specification Test Results

Assay ≥99,0%
Heavy Metals ≤0,001% (as Pb)
Insoluble Matter ≤0,01%
Loss on Drying 13,0 - 15,0% (150 °C)
Chloride ≤0,002%
Nitrate ≤0,001%
Iron <0,001%
Potassium ≤0,05%
Sodium ≤0,05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR