ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) ≥95%

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0215174225 0215174250 0215174280 0215174283 0215174290 0215174291
ICNA0215174225EA 132 GBP
ICNA0215174225 ICNA0215174250 ICNA0215174280 ICNA0215174283 ICNA0215174290 ICNA0215174291
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) ≥95%
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)

50 mg/ml, clear colourless to slightly yellow solution.

Formula: C₂₁H₂₆N₇Na₄O₁₇P₃
MDL Number: MFCD00036263
CAS Number: 2646-71-1

Order Now

Specification Test Results

Synonyms β-NADPH; TPNH; Coenzyme II reduced form tetrasodium salt; Dihydronicotinamide-adenine dinucleotide phosphate tetrasodium salt; Triphosphopyridine nucleotide reduced tetrasodium salt
Form White Powder
Storage Store at -0 °C, Desiccate, Protect from light, Store under nitrogen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR