Ethylene glycol ≥99%, clear, colourless liquid

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

0215108983 0215108991
ICNA0215108983EA 36.3 GBP
ICNA0215108983 ICNA0215108991
Ethylene glycol ≥99%, clear, colourless liquid
Ethylene glycol

Ethylene glycol is a synthetic liquid substance that absorbs water. It is odorless, but has a sweet taste.


  • Japan fire code: 4.3W
  • Explosion limit: 15,3%


Ethylene glycol is typically used in cyclocondensation reactions with aldehydes and ketones to form 1,3-dioxolanes.


Ethylene glycol has been used as a carrier fluid during the fabrication of shear thickening fluid (composed of nanosize silica particles). It has also been used to compose previtrification and vitrification solutions for the cryopreservation of mouse embryos.

Formula: HOCH₂CH₂OH
MDL Number: MFCD00002885
CAS Number: 107-21-1
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR