Yeast hydrolysate enzymatic from brewers yeast, MP Biomedicals
Supplier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0210330480 0210330405 0210330425
ICNA0210330480EA 2860 GBP
ICNA0210330480 ICNA0210330405 ICNA0210330425
Yeast hydrolysate enzymatic from brewers yeast, MP Biomedicals
Media for Microbiology Base Materials for Microbiology
Yeast hydrolysate enzymatic is an autolysed yeast extract produced from 100% brewers yeast. It is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.
  • Brown powder