Starch, soluble rice, powder_white

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Starch, Vitex® iodine indicator, Amylodextrin

0210295590 0210295591
ICNA0210295590EA 54.8 GBP
ICNA0210295590 ICNA0210295591
Starch, soluble rice, powder_white
Starch, soluble

Starch can be hydrolysed into simpler carbohydrates by acids, various enzymes, or a combination of the two and can be used as a bland-tasting filler, thickener, sweetner and in hydrogen production.


Starch can be hydrolysed into simpler carbohydrates by acids, various enzymes, or a combination of the two and can be used as a bland-tasting filler, thickener, sweetner and in hydrogen production.


Starch becomes soluble in water when heated.


Caution: Research Use Only (RUO).

Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
Merck Index: 13,08878

Order Now

Specification Test Results

Identity Test Passes
Source Rice


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR