Inosine 5'-monophosphate disodium salt hydrate ≥98%, white powder

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 5'-IMP 2Na hydrate, 5'-Inosinic acid disodium salt hydrate, Inosinic acid (disodium)(hydrate)(1:2:X), Inosine 5'-monophosphate disodium salt xhydrate, Disodium inosinate hydrate, Inosic acid disodium salt hydrate

0210205005 0210205010 0210205025 0210205080
ICNA0210205005EA 48.5 GBP
ICNA0210205005 ICNA0210205010 ICNA0210205025 ICNA0210205080
Inosine 5'-monophosphate disodium salt hydrate ≥98%, white powder
Inosine 5'-monophosphate disodium salt hydrate

IMP a nucleotide produced by the deamination of adenosine monophosphate (AMP); it is the precursor of AMP and GMP in purine biosynthesis and an intermediate in purine salvage and in purine degradation.


Inosine 5′-monophosphate (IMP) is used as a substrate to study the distribution, specificity and kinetics of inosine-5′-monophosphate dehydrogenase (IMPDH).


Inosine monophosphate, or inosinic acid, is the ribonucleotide of hypoxanthine and the first nucleotide formed during purine synthesis.


Soluble in water (5% aq soln).


Caution: Research Use Only (RUO).

Formula: C₁₀H₁₁N₄Na₂O₈P·xH₂O
MW: 392.17 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD00150372
CAS Number: 4691-65-0

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
UV/VIS: λ max 248 ± 2 nm
OD250/OD260 1.7 ± 0.2
OD280/OD260 0.3 ± 0.2
Moisture Repoer result
Solubility (5% aq soln) Passes

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR