Dextran, M.W. 200000 - 300000, white powder clinical grade

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0210151450 0210151480 0210151490 0210151491
ICNA0210151450EA 89.3 GBP
ICNA0210151450 ICNA0210151480 ICNA0210151490 ICNA0210151491
Dextran, M.W. 200000 - 300000, white powder clinical grade
Dextran

Dextran is a high molecular weight polymer of anhydroglucose. It is composed of approximately 95% alpha-D-(1-->6) linkages. The remaining a(1-->3) linkages account for the branching of dextran.
Dextrans are used in many applications as volume extenders, stabilizers, matrix components, binding platforms, lubricants and physical structure components.


  • Intrinsic viscosity: 0,4 to 0,5 (37 °C) for 1% solution


Dextrans are used in many applications as volume extenders, stabilizers, matrix components, binding platforms, lubricants and physical structure components.Use of dextrans as long and hydrophilic spacer arms improves the performance of immobilized proteins acting on macromolecules.Dextrans are used in many applications as platelet aggregants, plasma volume extenders, osmotic pressure regulators, stabilizers, organ separation media, matrix components, copolymers, microcarriers, binding agents, viscosity modifiers, antithrombotics, lubricants and physical structure components.[2] Dextrans may be used as long hydrophilic spacer arms to improve the performance (freedom of movement) of conjugated/bound proteins. Dextrans may be derivatized for use in biosensor systems. Dextran 150 may be used to study iron delivery to cells, cell membrane permeabilization and red blood cell aggregation


Caution: Research Use Only (RUO).

Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00130935
CAS Number: 9004-54-0

Order Now

Specification Test Results

Identity Test Passes
pH (1% aq soln) 4.0-7.0
Moisture ≤10%
Color by Colorimetric <0.15 abs @ 375 nm
Intrinsic Viscosity 0.4 - 0.5 (37°C)
Sulphated Ash ≤2.0%
Cold Water Solubility Passes

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR