Cysteaminium chloride ∼98%, white powder

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Aminoethanethiol hydrochloride, Cysteamine Hydrochloride, 2-Mercaptoethylamine hydrochloride, 2-MEA, β-Mercaptoethylamine hydrochloride, Mercaptoethylamine-HCl, H-Cys-NH2·HCl

0210099505 0210099525 0210099580
ICNA0210099505EA 39 GBP
ICNA0210099505 ICNA0210099525 ICNA0210099580
Cysteaminium chloride ∼98%, white powder
Cysteaminium chloride

β-Mercaptoethylamine (Cysteamine hydrochloride) is useful antioxidant. It is experimentally used as a radioprotective agent. It is also used to produce acute and chronic duodenal ulcers in rats and as an antidote to acetaminophen.


Soluble in water (50 mg/ml) clear, colorless to faint yellow, soluble in alcohol.


Caution: Research Use Only (RUO).

Formula: C₂H₇NS·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00012904
CAS Number: 156-57-0

Order Now

Specification Test Results

Identity Test Passes
Purity ~98%
FTIR Conforms to Standard
Melting Point 58 - 73 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR