Adenosine free base ≥98%, white powder

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside

0210019980 0210019905 0210019925 0210019991
ICNA0210019980EA 258 GBP
ICNA0210019980 ICNA0210019905 ICNA0210019925 ICNA0210019991
Adenosine free base ≥98%, white powder
Adenosine

Adenosine is a nucleotide resulting from the union of one molecule of sugar -- d-ribose -- with a base -- l-adenine.


Adenosine is active in cardiovascular disorders. It is indicated in cases of angina pectoris on account of its property of increasing the coronary output and in cases of hypertension on account of its vasodilator action. It is also active on the peripheral circulation, dilating the capillary vessels. It has also been recommended in the treatment of agranulocytosis. It induces apoptosis in human leukemia HL-60 cells and also has anti-arrhythmic properties.


Endogenous neurotransmitter at adenosine receptors. Cardioprotective effects may relate to activation of A1 adenosine receptors. The antiplatelet and anti -inflammatory actions of adenosine appear to be mediated via the A2 adenosine receptor. In contrast, adenosine appears to be a pro-inflammatory mediator in asthma and chronic obstructive pulmonary disease (COPD).


Fairly soluble in cold water, soluble in room temperature water, freely soluble in hot water; soluble in 1 N hydrochloric acid (50 mg/ml - clear and colourless); insoluble in alcohol.

Formula: C₁₀H₁₃N₅O₄
Storage Temperature: Freezer
MDL Number: MFCD00005752
CAS Number: 58-61-7
EINECS: 200-389-9

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
Melting Point 228 - 236 °C
UV/VIS: l max (1N HCl) 255 - 260 nm
OD250/OD260 0,80 - 0,90
OD280/OD260 0,17 - 0,24


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR