Aluminium oxide neutral, MP EcoChrom™ H15152

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0204654
ICNA0204654EA 0 GBP
ICNA0204654
Aluminium oxide neutral, MP EcoChrom™ H15152
Aluminium oxide neutral
Formula: Al₂O₃
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR