Sample concentrators, Reacti Vap™
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TS-18825 TS-18826
HYPETS-18825EA 901 GBP
HYPETS-18825 HYPETS-18826
Sample concentrators, Reacti Vap™
Block Heaters Sample Concentrators
Reacti-Vap Evaporators (9 port)
These evaporators are precision machined gassing manifolds. They provide simple, efficient evaporation by allowing the simultaneous or separate delivery of non-reactive pressurised gas to samples. They easily connect to single block Reacti-Therm™ modules.
Reacti-Vap III Evaporators (27 port)
These evaporators triple the number of samples to evaporate. Nine needles attach to each of the three individually regulated chambers. They easily connect to Reacti- Therm™ III modules.