Two-dimensional BioProcess Container™ system, Labtainer™ BPC bags
Supplier: THERMO BIOPROCESS DIVISION
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SH30709.02 SH30714.03 SH30712.04 SH30657.11 SH30657.13 SH30657.14 SH30657.15 SH30657.16 SH30658.11 SH30658.12 SH30658.13 SH30658.14 SH30658.15 SH30662.11 SH30658.16 SH30662.12 SH30662.14 SH30662.15 SH30662.16 SH30713.05 SH30714.05 SH30712.05 SH30713.01 SH30714.01 SH30714.02 SH30713.03 SH30712.02 SH30712.03 SH30713.04 SH30714.04 SH30709.01 SH30709.04 SH30709.05 SH30713.02 SH30709.03
HYCPSH30709.02EA 108 GBP
HYCPSH30709.02 HYCPSH30714.03 HYCPSH30712.04 HYCPSH30657.11 HYCPSH30657.13 HYCPSH30657.14 HYCPSH30657.15 HYCPSH30657.16 HYCPSH30658.11 HYCPSH30658.12 HYCPSH30658.13 HYCPSH30658.14 HYCPSH30658.15 HYCPSH30662.11 HYCPSH30658.16 HYCPSH30662.12 HYCPSH30662.14 HYCPSH30662.15 HYCPSH30662.16 HYCPSH30713.05 HYCPSH30714.05 HYCPSH30712.05 HYCPSH30713.01 HYCPSH30714.01 HYCPSH30714.02 HYCPSH30713.03 HYCPSH30712.02 HYCPSH30712.03 HYCPSH30713.04 HYCPSH30714.04 HYCPSH30709.01 HYCPSH30709.04 HYCPSH30709.05 HYCPSH30713.02 HYCPSH30709.03
Two-dimensional BioProcess Container™ system, Labtainer™ BPC bags
Bags Media Bags
BioProcess Container™ (BPC™) systems include two-dimensional single-use bags for liquid handling. Manufactured from medical-grade plastics (cGMP compliant) and configured with ports, tubing and fittings, BPC™ systems provide an efficient means for sterile liquid handling in research and production settings.
  • Film type: CX5-14 Single Web
  • Sterilisation method: Gamma irradiation