Stopcocks for carboys
Supplier: hünersdorff GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
520100 520700 520705 520105
216-5291EA 22.7 GBP
216-5291 216-5292 HUEN520705 HUEN520105
Stopcocks for carboys
Stopcocks
Made of fracture-proof and food-safe polyethylene with sleeve connection for a hose.
  • Very resistant to acids, alkalis and other chemicals
  • allow for extra-precise dosing