Water baths, insulated stainless steel
Supplier: HUBER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6055 6056 6057 6058 6059 6060 6215 6329 6330 6331
HUBE6055EA 2930 GBP
HUBE6055 HUBE6056 HUBE6057 HUBE6058 HUBE6059 HUBE6060 HUBE6215 HUBE6329 HUBE6330 HUBE6331
Water baths, insulated stainless steel
Baths Water Baths, Unstirred
These insulated stainless steel baths are available in 3 standard sizes and can be customised to suit requirements.
Can be fitted with inlet and outlet connections directly into the bath (direct temperature control) or into the double-wall for indirect temperature control. The drain is on the short side as standard but can be fitted on the long side on request.