Compact Recirculation Chiller/Circulation Chiller, Minichiller 800 and 1000 OLÉ
Supplier: HUBER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3079.0001.98 3080.0001.98
HUBE3079.0001.98EA 5.9 GBP
HUBE3079.0001.98 HUBE3080.0001.98
Compact Recirculation Chiller/Circulation Chiller, Minichiller 800 and 1000 OLÉ
Circulating Thermostats Closed Circulator Systems Cooling Circulators
The compact mini chillers are a cost-effective and environmentally friendly cooling solution for numerous laboratory applications.
  • Environmentally friendly, economical cooling solution
  • Robust stainless steel construction
  • Illuminated level indicator
  • USB and RS232 interface
  • High efficiency
  • Continuous operation with warning functions
  • Large, bright OLED display
  • Precise temperature control
  • Easy filling and emptying
  • Easy operation with menu navigation
A calculation for water and waste water costs shows that a mini chiller can save up to 48000 liters ofMore Product Information