Sharpsafe® Ecological 5th Generation Recycled Needle Containers
Supplier: Hospidex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SHA-51202431 SHA-51405610 SHA-51405720 SHA-51455610 SHA-51455720 SHA-51502430 SHA-51502610 SHA-51702610 SHA-51703610 SHA-51704610 SHA-51707610 SHA-51732610 SHA-51733610 SHA-51902610 SHA-51912613 SHA-51922613 SHA-50502610 SHA-51005610 SHA-51005720 SHA-51105610 SHA-51105720 SHA-51116610 SHA-51152610 SHA-51172610 SHA-51602610 SHA-51602720 SHA-51802431 SHA-51005410 SHA-51005420 SHA-51005430 SHA-51105430 SHA-51152430 SHA-51172430 SHA-51202411 SHA-51405430 SHA-51455430 SHA-51802411
HUARSHA-51202431EA 548 GBP
HUARSHA-51202431 HUARSHA-51405610 HUARSHA-51405720 HUARSHA-51455610 HUARSHA-51455720 HUARSHA-51502430 HUARSHA-51502610 HUARSHA-51702610 HUARSHA-51703610 HUARSHA-51704610 HUARSHA-51707610 HUARSHA-51732610 HUARSHA-51733610 HUARSHA-51902610 HUARSHA-51912613 HUARSHA-51922613 HUARSHA-50502610 HUARSHA-51005610 HUARSHA-51005720 HUARSHA-51105610 HUARSHA-51105720 HUARSHA-51116610 HUARSHA-51152610 HUARSHA-51172610 HUARSHA-51602610 HUARSHA-51602720 HUARSHA-51802431 HUARSHA-51005410 HUARSHA-51005420 HUARSHA-51005430 HUARSHA-51105430 HUARSHA-51152430 HUARSHA-51172430 HUARSHA-51202411 HUARSHA-51405430 HUARSHA-51455430 HUARSHA-51802411
Sharpsafe® Ecological 5th Generation Recycled Needle Containers
Bins Sharps Containers
Sharpsafe®, historic pioneer in protecting against the risk of sharps injuries and sustainability in medical waste packaging management, presents the recycled 5th generation needle containers.
  • Increased efficiency to reduce sharps injuries
  • Reduced carbon footprint (recycled plastic, made in Europe)
  • Easy 4 click assembly with intuitive one-handed disconnection
  • Clear fill limit line
  • Temporary and permanent closing
  • Puncture-resistant
  • Clear labelling in national language
The Sharpsafe® range of sharps containers is a sharps management system thatMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001 (EMS 74142)