VWR® Degassers, PTFE AF
Supplier: COLLECTION PREFIX HPLC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1-6353A 0003-6623 0001-6624 0003-6626 0003-6625
903-0134EA 0 GBP
903-0134 903-0156 903-0157 HPLC0003-6626 HPLC0003-6625
VWR® Degassers, PTFE AF
Degassers
On-Line degassing has been shown to be more effective at removing dissolved gas from mobile phases than sonication or helium sparging. Up to five solvent lines can be degassed simultaneously by one unit. The extremely low volume of each PTFE AF channel (480 μl) offers very quick equilibration and very short start-up times.
  • Considerably shorter equilibration/re-equilibration times
  • Very easy to prime
  • Short vacuum pull-down times, typically 3 minutes
  • PTFE AF has the ability to remove pre-pump bubbles
ZHCR® (Zero Hysteresis Constant Run) stepper motorMore Product Information