Ice Cubers, Self Contained, IM
Supplier: HOSHIZAKI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
IM-100CNE-HC21 IM-100CNE-HC
HOSHIM-100CNE-HC21EA 0 GBP
HOSHIM-100CNE-HC21 HOSHIM-100CNE-HC
Ice Cubers, Self Contained, IM
Ice Makers
Self contained cube ice makers producing high-quality cube ice. Meeting even the strictest hygiene requirements, this true allrounder offers many application possibilities in segments including the foodservice and medical sector.
  • Uses refrigerant R290, this natural refrigerant is not only climate-friendly, but also boosts the machine, when it comes to performance and production capacity
  • The exterior is made of stainless steel
  • The closed water circuit provides maximum protection against any type of contamination during the entire cubing process
  • Magnetic
More Product Information