Ice maker, CM series
Supplier: HOSHIZAKI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CM140-KE
HOSHCM140-KEEA 0 GBP
HOSHCM140-KE
Ice maker, CM series
Ice Makers
The CM series produce cubelet ice that is easy to maintain and clean. It is used in the food, beverage and healthcare sectors.
  • Automatic flush function: regular flushing to keep the ice making system clean
  • One-piece bin moulding: round corners are hygienic and easy to keep clean
  • Integrated door handle design: easy to open and to clean
  • Controller board with operation status display: ensuring product durability and low maintenance costs