Erbium AAS standards, (CRMs), Fluka™
Supplier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E4025-100ML E4025-500ML
HONCE4025-100MLEA 81.6 GBP
HONCE4025-100ML HONCE4025-500ML
Erbium AAS standards, (CRMs), Fluka™
Standards Cationic Standards Erbium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).