Ammonium (N) standard for ion chromatography, (CRM), Fluka™
Supplier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E3002-100ML
HONCE3002-100MLEA 69.5 GBP
HONCE3002-100ML
Ammonium (N) standard for ion chromatography, (CRM), Fluka™
Standards Cationic Standards Ammonium Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).