Initial calibration verification standard 2 Hg for ICP
Supplier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E2181-100ML
HONCE2181-100MLEA 337 GBP
HONCE2181-100ML
Initial calibration verification standard 2 Hg for ICP
Standards Cationic Standards Multi-Element Standards
Certified Reference Material (CRM).
10 mg/L each of Sn, Ti in HNO₃ 2%