Dysprosium ICP & ICP-MS standards, (CRMs), Fluka™
Supplier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E1315-100ML E1383-100ML
HONCE1315-100MLEA 143 GBP
HONCE1315-100ML HONCE1383-100ML
Dysprosium ICP & ICP-MS standards, (CRMs), Fluka™
Standards Cationic Standards Dysprosium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Order Now
SPECIFICATIONS
Environmentally Preferable