Chromium ICP standards, (CRMs), Fluka™
Supplier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
E1015-100ML E1104-100ML E1142-100ML
HONCE1015-100MLEA 101 GBP
HONCE1015-100ML HONCE1104-100ML HONCE1142-100ML
Chromium ICP standards, (CRMs), Fluka™
Standards Cationic Standards Chromium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).