Methanol ACS, gradient grade for HPLC, Burdick & Jackson™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

65727-2.5L 65727-7L-RC 65727-45L 65727-1000L 65727-30L-RC 65727-395L
HONC65727-2.5LEA 135 GBP
HONC65727-2.5L HONC65727-7L-RC HONC65727-45L HONC65727-1000L HONC65727-30L-RC HONC65727-395L
Methanol ACS, gradient grade for HPLC, Burdick & Jackson™
Methanol
Formula: CH₃OH
Boiling Pt: 64…65 °C (1.013 hPa)
Melting Pt: –98 °C
Density: 0.79 g/cm3 (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance complying
Assay (GC) Min. 99.95 %
Non-volatile matter Max. 0.0001 %
Density (D 20/4) 0.7900 - 0.7925
Refractive index (n 20/D) 1.328 - 1.330
Water (Karl Fischer) Max. 0.02 %
Acidity Max. 0.0002 meq/g
Alkalinity Max. 0.0002 meq/g
Heavy metals (as Pb) Max. 2 ppm
Acetaldehyde Max. 0.001 %
Acetone (GC) Max. 0.001 %
Formaldehyde Max. 0.001 %
KMnO₄ reducing matter complying
Reaction against H₂SO₄ complying
Mixable with H₂O complying
Transmittance at 210 nm Min. 33 %
Transmittance at 220 nm Min. 60 %
Transmittance at 235 nm Min. 85 %
Transmittance from 260 nm Min. 98 %
Absorbance complying
HPLC Gradient at 230 nm Max. 2 mAU
HPLC Gradient at 254 nm Max. 2 mAU
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
APHA Max. 10
Suitability for residue analysis Max. 3 ng/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR