Tetrahydrofuran, Burdick & Jackson™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

65529-177KG
HONC65529-177KGEA 7030 GBP
HONC65529-177KG
Tetrahydrofuran, Burdick & Jackson™
Tetrahydrofuran
Formula: C₄H₈O
Boiling Pt: 65 °C
Density: 0.89 g/cm3
Flash Pt: –21 °C
MDL Number: MFCD00005356
CAS Number: 109-99-9
UN: 2056
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.5 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.05 %
Peroxides (as H₂O₂) Max. 0.005 %
Assay BHT (GC) 100 - 300 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR