Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

34860-7L 34860-18L 34860-45L 34860-2.5L 34860-4X2.5L 34860-1L 34860-6X1L 34860-2L 34860-4X2L 34860-30L
34860-7LEA 380 GBP
34860-7L 34860-18L-R 34860-45L-R 34860-2.5L 34860-4X2.5L 34860-1L 34860-6X1L 34860-2L 34860-4X2L HONC34860-30L
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Methanol

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: CH₃OH
Boiling Pt: 64…65 °C (1.013 hPa)
Melting Pt: –98 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.03 %
Absorbance at 400 nm Max. 0.01
Absorbance 280 nm Max. 0.01
Absorbance at 260 nm Max. 0.04
Absobance at 254 nm Max. 0.02
Absorbance at 240 nm Max. 0.07
Absorbance at 235 nm Max. 0.10
Absorbance at 230 nm Max. 0.15
Absorbance at 220 nm Max. 0.30
Absorbance at 210 nm Max. 0.60
Absorbance at 205 nm Max. 1.00
Absorbance (JP)(Val.Ref) complying
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
APHA Max. 10
Vapor Pressure 410 mmHg ( 50 °C)
Vapor Density 1.11 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR