Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


Stabilisation: Stabilised with ethanol 0.6 - 1.0%

32211-1L 32211-2.5L 32211-4X2.5L 32211-6X1L 32211-18L 32211-195L 32211-7L
32211-1LEA 78.8 GBP
32211-1L 32211-2.5L 32211-4X2.5L 32211-6X1L HONC32211-18L HONC32211-195L HONC32211-7L
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Chloroform
Formula: CHCl₃
Boiling Pt: 60…62 °C (1.013 hPa)
Melting Pt: –63 °C
Density: 1.48 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (GC) complying
Assay (GC) 99.0 - 99.4 %
Boiling range 60 - 62 °C
Density (D 20/20) 1.476 - 1.483
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as HCl) Max. 0.00005 %
Acid and Phosgene complying
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000005 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Chloride (Cl) Max. 0.0001 %
Free chlorine (Cl) Max. 0.00001 %
Aldehydes, Ketones (as CH₃CocH₃) Max. 0.005 %
Carbonyl compounds (as CO) Max. 0.005 %
Dichloromethane (GC) Max. 0.01 %
Ethanol (GC) 0.6 - 1.0 Gew.%
Tetrachloroethene (GC) Max. 0.01 %
Tetrachloromethane (GC) Max. 0.01 %
Trichloroethene (GC) Max. 0.01 %
Other Impurities (GC) Max. 0.1 %
Reaction against H₂SO₄ complying
Appearance of the substance complying
Suitability for determinations with dithizone (ISO) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR